Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127714.2 | NP_001121186.1 | 3 | 1228 | |
NM_024503.4 | NP_078779.2 | 4 | 1308 | ||
XM_011541884.2 | XP_011540186.1 | 5 | 1240 | ||
XM_011541885.2 | XP_011540187.1 | 4 | 1520 | ||
XM_011541886.2 | XP_011540188.1 | 4 | 1520 | ||
XM_011541887.2 | XP_011540189.1 | 4 | 1520 | ||
XM_017001992.1 | XP_016857481.1 | 5 | 2487 | ||
XM_017001993.1 | XP_016857482.1 | 3 | 1440 | ||
XM_017001994.1 | XP_016857483.1 | 4 | 1520 | ||
GenBank mRNA | AB046775.3 | - | 3 | 1228 | |
AF278765.1 | - | 4 | 1101 | ||
BC152563.1 | - | 3 | 1228 | 8860523 |