Gene Transcripts

Gene Symbol
SLC35B4
Entrez Gene ID
84912
Gene Name
solute carrier family 35 member B4
Gene Aliases
YEA, YEA4
Chromosome Location
Chr. 7: 134289337 - 134320307 on Build GRCh38
UniGene ID
Hs.490181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032826.4 NP_116215.1 10 1640
XM_011516645.2 XP_011514947.1 10 1408
GenBank mRNA AB052892.1 - 10 1616
AJ315497.1 - 10 1472
AK075368.1 - 10 1445
AK222874.1 - 10 1489

View Full Product Details