Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000577.4 | NP_000568.1 | 5 | 1600 | |
NM_001318914.1 | NP_001305843.1 | 7 | 1834 | ||
NM_173841.2 | NP_776213.1 | 6 | 1663 | ||
NM_173842.2 | NP_776214.1 | 4 | 1592 | ||
NM_173843.2 | NP_776215.1 | 6 | 1771 | ||
XM_005263661.4 | XP_005263718.1 | 5 | 1619 | ||
XM_011511121.1 | XP_011509423.1 | 9 | 2122 | ||
GenBank mRNA | BC009745.2 | - | 5 | 1505 | |
X52015.1 | - | 4 | 1551 | ||
X53296.1 | - | 4 | 1536 |