Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143838.2 | NP_001137310.1 | 2 | 206 | |
NM_001284509.1 | NP_001271438.1 | 2 | 206 | ||
NM_177550.4 | NP_808218.1 | 2 | 206 | ||
XM_011523795.2 | XP_011522097.1 | 2 | 196 | ||
GenBank mRNA | AK127797.1 | - | 2 | 154 | |
AK172785.1 | - | 2 | 151 | ||
AY151833.1 | - | 2 | 130 | ||
BC104795.1 | - | 2 | 162 | 8143798 | |
BC112151.1 | - | 2 | 162 | 8327619 | |
BC143689.1 | - | 2 | 162 | 9052203 | |
DA187094.1 | - | 2 | 206 |