Gene Transcripts

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
Chromosome Location
Chr. 16: 31108294 - 31117651 on Build GRCh38
UniGene ID
Hs.513520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122957.2 NP_001116429.1 11 1326
NM_001271926.1 NP_001258855.1 10 1236
NM_005881.3 NP_005872.2 11 1326
XM_017022859.1 XP_016878348.1 11 1239
GenBank mRNA AF026548.1 - 11 1236
AK130145.1 - 11 1206
BC007363.2 - 11 1209 3630050
BC009872.2 - 10 1099 3944953
CR542093.1 - 10 963

View Full Product Details