Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007794.1 | NP_001007795.1 | 2 | 411 | |
NM_006090.3 | NP_006081.1 | 2 | 373 | ||
XM_006710285.3 | XP_006710348.1 | 2 | 365 | ||
XM_011540496.1 | XP_011538798.1 | 2 | 373 | ||
GenBank mRNA | AF068302.1 | - | 2 | 331 | |
AF138862.1 | - | 1 | 241 | ||
AK023381.1 | - | 2 | 360 | ||
AL833102.1 | - | 2 | 384 | ||
BC032610.2 | - | 3 | 373 | 5554219 | |
BC049196.1 | - | 2 | 411 | 5226538 | |
BX487092.1 | - | 2 | 373 |