Gene Transcripts

Gene Symbol
SULT1B1
Entrez Gene ID
27284
Gene Name
sulfotransferase family 1B member 1
Gene Aliases
ST1B1, ST1B2, SULT1B2
Chromosome Location
Chr. 4: 69726848 - 69760712 on Build GRCh38
UniGene ID
Hs.129742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014465.3 NP_055280.2 7 989
XM_005265676.3 XP_005265733.1 8 904
XM_005265677.3 XP_005265734.1 8 1104
XM_005265678.3 XP_005265735.1 7 857
XM_011531873.2 XP_011530175.1 6 728
GenBank mRNA BC010895.1 - 7 991
D89479.1 - 6 715
U95726.1 - 6 706

View Full Product Details