Gene Transcripts

Gene Symbol
RRM2B
Entrez Gene ID
50484
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
MTDPS8A, MTDPS8B, P53R2
Chromosome Location
Chr. 8: 102204501 - 102239118 on Build GRCh38
UniGene ID
Hs.512592
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172477.1 NP_001165948.1 2 329
NM_015713.4 NP_056528.2 2 348
GenBank mRNA AB036063.1 - 2 348
AB163438.1 - 2 104
AB166671.1 - 2 104
AK001965.1 - 2 143
AK304354.1 - 2 273
AL137348.1 - 2 150
BC108261.1 - 2 160 6143754
BC117496.1 - 2 185 40126047
BC130628.1 - 2 185 40146659
CR627376.1 - 2 273
DC308409.1 - 2 329

View Full Product Details