Gene Transcripts

Gene Symbol
AKR1E2
Entrez Gene ID
83592
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
AKR1CL2, AKRDC1, LoopADR, hTSP, htAKR
Chromosome Location
Chr. 10: 4824554 - 4908421 on Build GRCh38
UniGene ID
Hs.657944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040177.2 NP_001035267.1 4 569
NM_001271021.1 NP_001257950.1 4 569
NM_001271025.1 NP_001257954.1 4 569
NR_073125.1 4 569
NR_073126.1 4 483
NR_073127.1 4 569
XM_011519715.2 XP_011518017.1 4 866
XM_011519718.2 XP_011518020.1 4 866
XM_011519719.2 XP_011518021.1 4 866
XM_011519720.2 XP_011518022.1 4 865
XM_011519722.2 XP_011518024.1 4 866
XM_011519724.2 XP_011518026.1 4 865
XM_011519725.2 XP_011518027.1 4 863
XM_011519729.2 XP_011518031.1 4 863
XM_017016743.1 XP_016872232.1 4 867
XM_017016744.1 XP_016872233.1 4 866
XM_017016745.1 XP_016872234.1 4 652
GenBank mRNA AB040820.1 - 4 450
AB040821.1 - 4 450
AB040822.1 - 4 450
AB055603.1 - 4 421
AF263242.1 - 4 441
AK074211.1 - 4 483
AK302155.1 - 3 374
BC002862.2 - 4 512 3941289
DA006578.1 - 4 536

View Full Product Details