Gene Transcripts

Gene Symbol
TRPV3
Entrez Gene ID
162514
Gene Name
transient receptor potential cation channel subfamily V member 3
Gene Aliases
FNEPPK2, OLMS, VRL3
Chromosome Location
Chr. 17: 3510502 - 3557995 on Build GRCh38
UniGene ID
Hs.446255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258205.1 NP_001245134.1 18 2926
NM_145068.3 NP_659505.1 18 2923
GenBank mRNA AF514998.1 - 18 2665
AK127330.1 - 6 922
AK127726.1 - 12 1875
AY118268.1 - 18 2736
BX537539.1 - 12 1861

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
Chromosome Location
Chr. 17: 3440011 - 3513852 on Build GRCh38
UniGene ID
Hs.351068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321336.1 NP_001308265.1 1 164
NM_001321337.1 NP_001308266.1 1 164
GenBank mRNA AK295299.1 - 1 164

View Full Product Details