Gene Transcripts

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
Chromosome Location
Chr. 4: 107931549 - 107953461 on Build GRCh38
UniGene ID
Hs.109087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183075.2 NP_898898.1 3 1304
XM_005262717.2 XP_005262774.1 3 1370
XM_005262720.2 XP_005262777.1 2 680
GenBank mRNA AK290002.1 - 3 1309
AK298989.1 - 2 226
AK303284.1 - 4 1109
AK304724.1 - 4 1182
AY343323.1 - 3 1260
BC132767.1 - 3 1269 40146789
BC136483.1 - 3 1269 9020472

View Full Product Details