Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002318.2 | NP_002309.1 | 3 | 832 | |
GenBank mRNA | AK222477.1 | - | 3 | 743 | |
AK312266.1 | - | 3 | 744 | ||
BC000594.2 | - | 3 | 702 | 3347512 | |
BC020481.1 | - | 3 | 752 | ||
KF928961.1 | - | 2 | 493 | ||
KF975332.1 | - | 2 | 505 | ||
U89942.1 | - | 3 | 740 |