Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003036.3 | NP_003027.1 | 3 | 1242 | |
XM_005244775.3 | XP_005244832.1 | 3 | 1212 | ||
XM_005244776.4 | XP_005244833.1 | 3 | 4355 | ||
XM_017002128.1 | XP_016857617.1 | 3 | 1019 | ||
GenBank mRNA | X15218.1 | - | 3 | 1242 |