Gene Transcripts

Gene Symbol
SLC2A4RG
Entrez Gene ID
56731
Gene Name
SLC2A4 regulator
Gene Aliases
GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19
Chromosome Location
Chr. 20: 63739858 - 63744050 on Build GRCh38
UniGene ID
Hs.435126
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020062.3 NP_064446.2 8 1961
GenBank mRNA AV659783.2 - 1 297

View Full Product Details