Gene Transcripts
- Gene Symbol
- ZNF57
- Entrez Gene ID
- 126295
- Gene Name
- zinc finger protein 57
- Gene Aliases
- ZNF424
- Chromosome Location
- Chr. 19: 2900898 - 2918646 on Build GRCh38
- UniGene ID
- Hs.591378
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028974.1 |
- |
3 |
239 |
|
View Full Product Details