Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001093.3 | NP_001084.3 | 3 | 859 | |
XM_005253876.4 | XP_005253933.1 | 4 | 969 | ||
XM_006719367.3 | XP_006719430.1 | 3 | 490 | ||
XM_011538259.2 | XP_011536561.1 | 4 | 1607 | ||
XM_011538263.2 | XP_011536565.1 | 4 | 1607 | ||
XM_011538264.2 | XP_011536566.1 | 3 | 944 | ||
XM_011538265.2 | XP_011536567.1 | 4 | 1609 | ||
XM_017019252.1 | XP_016874741.1 | 3 | 474 | ||
GenBank mRNA | AJ575431.2 | - | 3 | 850 | |
AJ575592.3 | - | 3 | 850 | ||
AY382667.1 | - | 4 | 969 | ||
DQ493870.1 | - | 3 | 850 | ||
U89344.1 | - | 3 | 847 |