Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
6566
Gene Name
solute carrier family 16 member 1
Gene Aliases
HHF7, MCT, MCT1, MCT1D
Chromosome Location
Chr. 1: 112911847 - 112956353 on Build GRCh38
UniGene ID
Hs.75231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166496.1 NP_001159968.1 5 3476
NM_003051.3 NP_003042.3 5 3007
XM_011542026.2 XP_011540328.1 5 2740
XM_011542027.2 XP_011540329.1 5 2750
GenBank mRNA AJ438942.2 - 1 1346
AK000641.1 - 2 1892
AL162079.1 - 5 2837
BC026317.1 - 5 2850 4811581

View Full Product Details