Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004790.4 | NP_004781.2 | 10 | 1979 | |
NM_153276.2 | NP_695008.1 | 10 | 1940 | ||
NM_153277.2 | NP_695009.1 | 10 | 1808 | ||
NM_153278.2 | NP_695010.1 | 10 | 1847 | ||
XM_017018562.1 | XP_016874051.1 | 10 | 1891 | ||
GenBank mRNA | AB009697.1 | - | 10 | 1913 | |
AB009698.1 | - | 10 | 1871 | ||
AF057039.2 | - | 10 | 1851 | ||
AF097490.1 | - | 10 | 1607 | ||
AF104038.1 | - | 10 | 1848 | ||
AF124373.1 | - | 10 | 1864 | ||
AJ251529.1 | - | 10 | 1475 | ||
AJ271205.1 | - | 10 | 1514 | ||
AK055764.1 | - | 11 | 1853 | ||
AK091879.1 | - | 10 | 1865 | ||
AK298374.1 | - | 11 | 1750 | ||
BC033682.1 | - | 10 | 1900 | 5190748 |