Gene Transcripts

Gene Symbol
LPO
Entrez Gene ID
4025
Gene Name
lactoperoxidase
Gene Aliases
SPO
Chromosome Location
Chr. 17: 58238426 - 58268518 on Build GRCh38
UniGene ID
Hs.234742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160102.1 NP_001153574.1 11 2618
NM_006151.2 NP_006142.1 13 2867
NR_027647.1 12 2779
XM_011524808.2 XP_011523110.1 11 2329
XM_011524810.2 XP_011523112.1 7 2342
GenBank mRNA BX538344.1 - 9 1938
BX648309.1 - 13 2705
M58151.1 - 5 1388
U39573.1 - 13 2632

View Full Product Details