Gene Transcripts

Gene Symbol
TMX1
Entrez Gene ID
81542
Gene Name
thioredoxin related transmembrane protein 1
Gene Aliases
PDIA11, TMX, TXNDC, TXNDC1
Chromosome Location
Chr. 14: 51240168 - 51257655 on Build GRCh38
UniGene ID
Hs.125221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030755.4 NP_110382.3 4 541
GenBank mRNA AB048246.1 - 4 533
AK075395.1 - 4 448
AK303134.1 - 4 531
AK312905.1 - 4 419
AL080080.1 - 4 439
AY358640.1 - 4 428
BC036460.2 - 4 491 4814497
BC056874.1 - 4 430 5740203
DA203669.1 - 4 541

View Full Product Details