Gene Transcripts
- Gene Symbol
- SMS
- Entrez Gene ID
- 6611
- Gene Name
- spermine synthase
- Gene Aliases
- MRSR, SPMSY, SRS, SpS
- Chromosome Location
- Chr. X: 21940573 - 21994837 on Build GRCh38
- UniGene ID
- Hs.724874
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258423.1 |
NP_001245352.1
|
5, 6 |
837 |
|
NM_004595.4 |
NP_004586.2
|
7, 8 |
996 |
|
XM_005274582.2 |
XP_005274639.1
|
7, 8 |
1062 |
|
XM_011545568.2 |
XP_011543870.1
|
7, 8 |
856 |
|
XM_017029753.1 |
XP_016885242.1
|
7, 8 |
836 |
|
XM_017029754.1 |
XP_016885243.1
|
7, 8 |
803 |
|
XM_017029755.1 |
XP_016885244.1
|
7, 8 |
894 |
|
GenBank mRNA |
AD001528.1 |
- |
7, 8 |
845 |
|
AK293455.1 |
- |
6, 7 |
740 |
|
AK313834.1 |
- |
7, 8 |
828 |
|
BC009898.2 |
- |
7, 8 |
819 |
3506022 |
BC085621.1 |
- |
7, 8 |
788 |
5540463 |
Z49099.1 |
- |
7, 8 |
772 |
|
View Full Product Details