Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098511.2 | NP_001091981.1 | 3 | 484 | |
NM_001243952.1 | NP_001230881.1 | 4 | 687 | ||
NM_001243953.1 | NP_001230882.1 | 3 | 484 | ||
NM_004520.4 | NP_004511.2 | 3 | 484 | ||
GenBank mRNA | AK297347.1 | - | 3 | 230 | |
AK302270.1 | - | 3 | 178 | ||
AK307880.1 | - | 3 | 194 | ||
AK315866.1 | - | 3 | 168 | ||
BC031828.2 | - | 2 | 109 | 4932771 | |
DB452624.1 | - | 3 | 403 | ||
EF560716.1 | - | 3 | 195 | ||
EF560728.1 | - | 3 | 189 | ||
Y08319.1 | - | 2 | 110 |