Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006835.2 | NP_006826.1 | 7 | 1508 | |
XM_011531555.1 | XP_011529857.1 | 5 | 1333 | ||
GenBank mRNA | AB209766.1 | - | 7 | 1510 | |
AF135162.1 | - | 7 | 1163 | ||
AK222931.1 | - | 6 | 960 | ||
AK301111.1 | - | 6 | 1369 | ||
AK312632.1 | - | 9 | 1378 | ||
BC000420.2 | - | 7 | 1507 | 2964432 | |
BC004975.1 | - | 7 | 1507 | 2957878 | |
BT020019.1 | - | 6 | 964 | ||
CR541783.1 | - | 6 | 964 | ||
D50310.1 | - | 6 | 964 |