Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001215.3 | NP_001206.2 | 3 | 448 | |
NM_001270500.1 | NP_001257429.1 | 3 | 448 | ||
NM_001270501.1 | NP_001257430.1 | 2 | 268 | ||
XM_011542083.2 | XP_011540385.1 | 3 | 443 | ||
XM_011542084.2 | XP_011540386.1 | 3 | 496 | ||
GenBank mRNA | AB102863.1 | - | 3 | 395 | |
AB103091.1 | - | 2 | 215 | ||
AK300652.1 | - | 3 | 412 | ||
BP333541.1 | - | 3 | 447 | ||
BP334640.1 | - | 3 | 418 | ||
M57892.1 | - | 3 | 401 |