Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002533.3 | NP_002524.2 | 6 | 581 | |
NM_206840.2 | NP_996671.1 | 5 | 507 | ||
XM_011544196.1 | XP_011542498.1 | 6 | 564 | ||
XM_011544198.1 | XP_011542500.1 | 5 | 519 | ||
XM_011544199.1 | XP_011542501.1 | 5 | 473 | ||
XM_011544200.1 | XP_011542502.1 | 4 | 447 | ||
XM_017001378.1 | XP_016856867.1 | 7 | 673 | ||
XM_017001379.1 | XP_016856868.1 | 7 | 673 | ||
XM_017001380.1 | XP_016856869.1 | 7 | 673 | ||
XM_017001381.1 | XP_016856870.1 | 7 | 670 | ||
XM_017001382.1 | XP_016856871.1 | 6 | 603 | ||
XM_017001383.1 | XP_016856872.1 | 6 | 603 | ||
XM_017001384.1 | XP_016856873.1 | 7 | 673 | ||
XM_017001385.1 | XP_016856874.1 | 7 | 673 | ||
XM_017001386.1 | XP_016856875.1 | 4 | 447 | ||
GenBank mRNA | AK297070.1 | - | 6 | 540 | |
AK298826.1 | - | 5 | 459 | ||
AW997091.1 | - | 3 | 326 | ||
BC005897.1 | - | 6 | 540 | ||
BQ883523.1 | - | 1 | 101 | ||
U68140.1 | - | 6 | 575 | ||
U78772.1 | - | 5 | 488 |