Gene Transcripts

Gene Symbol
CDK5R2
Entrez Gene ID
8941
Gene Name
cyclin dependent kinase 5 regulatory subunit 2
Gene Aliases
NCK5AI, P39, p39nck5ai
Chromosome Location
Chr. 2: 218959628 - 218962155 on Build GRCh38
UniGene ID
Hs.158460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003936.4 NP_003927.1 1 1278
GenBank mRNA BC041771.1 - 1 1251
BT007437.1 - 1 269
U34051.1 - 1 1154

View Full Product Details