Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004252.4 | NP_004243.1 | 6 | 1289 | |
GenBank mRNA | AF015926.1 | - | 6 | 1266 | |
AF036241.1 | - | 6 | 1224 | ||
AK026581.1 | - | 6 | 1272 | ||
AK094467.1 | - | 5 | 935 | ||
AK128474.1 | - | 7 | 1238 | ||
AK312498.1 | - | 6 | 1305 | ||
BC001443.2 | - | 6 | 1242 | 3138881 | |
BC003361.1 | - | 6 | 1259 | 2900516 | |
BC011777.1 | - | 6 | 1258 | 3640162 | |
BC049220.1 | - | 6 | 1265 | ||
BC053350.1 | - | 6 | 1249 | 5218414 | |
BX648303.1 | - | 4 | 2948 |