Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011554.2 | NP_001011554.1 | 8 | 1037 | |
NM_001193339.1 | NP_001180268.1 | 6 | 851 | ||
NM_001193340.1 | NP_001180269.1 | 8 | 1037 | ||
NM_001193342.1 | NP_001180271.1 | 7 | 828 | ||
NM_022829.5 | NP_073740.2 | 7 | 1001 | ||
GenBank mRNA | AF154121.1 | - | 7 | 980 | |
AK056713.1 | - | 7 | 999 | ||
AK096658.1 | - | 8 | 1037 | ||
AK293902.1 | - | 7 | 817 | ||
AK295748.1 | - | 6 | 849 | ||
AK310960.1 | - | 6 | 1011 | ||
AK315131.1 | - | 7 | 999 | ||
AY072810.3 | - | 7 | 980 | ||
BC035966.1 | - | 7 | 999 | 4609776 | |
BM474324.1 | - | 432 | |||
DA272524.1 | - | 2 | 178 |