Gene Transcripts

Gene Symbol
RAB35
Entrez Gene ID
11021
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
H-ray, RAB1C, RAY
Chromosome Location
Chr. 12: 120095094 - 120116839 on Build GRCh38
UniGene ID
Hs.524788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167606.1 NP_001161078.1 3 313
NM_006861.6 NP_006852.1 3 313
XM_011537779.1 XP_011536081.1 3 250
XM_017018723.1 XP_016874212.1 3 215
GenBank mRNA AF498960.1 - 3 124
AK125543.1 - 4 1088
AK300127.1 - 3 231
AK304620.1 - 3 220
AK312538.1 - 3 223
BC015931.2 - 3 228 3907209
BQ233709.1 - 3 269
BT020024.1 - 3 124
CR536486.1 - 3 124
CR541683.1 - 3 124
DC349830.1 - 3 310
DC370610.1 - 3 313
DC408044.1 - 3 313
X79781.1 - 3 205

View Full Product Details