Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
22931
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
RAB18LI1, WARBM3
Chromosome Location
Chr. 10: 27504174 - 27542237 on Build GRCh38
UniGene ID
Hs.406799
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256410.1 NP_001243339.1 2, 3 319
NM_001256411.1 NP_001243340.1 2, 3 319
NM_001256412.1 NP_001243341.1 2, 3 319
NM_021252.4 NP_067075.1 2, 3 319
GenBank mRNA AF087860.1 - 2, 3 177
AF136974.1 - 2, 3 233
AF137372.1 - 2, 3 123
AF498950.1 - 2, 3 123
AK001555.1 - 2, 3 123
AK223153.1 - 2, 3 146
AK295443.1 - 2, 3 189
AK297629.1 - 2, 3 189
AL136734.1 - 2, 3 172
AY574034.1 - 2, 3 208
BC015014.2 - 2, 3 173 3922029
BC029350.1 - 2, 3 189 4819866
BT009840.1 - 2, 3 123
CN357871.1 - 2, 3 388
CR533455.1 - 2, 3 123

View Full Product Details