Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005173.3 | NP_005164.2 | 17 | 2739 | |
NM_174953.2 | NP_777613.1 | 17 | 2739 | ||
NM_174954.2 | NP_777614.1 | 17 | 2739 | ||
NM_174955.2 | NP_777615.1 | 17 | 2739 | ||
NM_174956.2 | NP_777616.1 | 17 | 2739 | ||
NM_174957.2 | NP_777617.1 | 17 | 2739 | ||
NM_174958.2 | NP_777618.1 | 17 | 2739 | ||
XM_011523881.2 | XP_011522183.1 | 17 | 2739 | ||
XM_011523882.2 | XP_011522184.1 | 17 | 2739 | ||
XM_011523884.2 | XP_011522186.1 | 17 | 2739 | ||
XM_011523885.1 | XP_011522187.1 | 17 | 2739 | ||
XM_011523888.2 | XP_011522190.1 | 17 | 2739 | ||
XM_011523889.1 | XP_011522191.1 | 17 | 2739 | ||
XM_017024692.1 | XP_016880181.1 | 17 | 2739 | ||
XM_017024693.1 | XP_016880182.1 | 15 | 2365 | ||
GenBank mRNA | AF068220.1 | - | 17 | 2690 | |
AF068221.1 | - | 5 | 967 | ||
AK292716.1 | - | 17 | 2739 | ||
BC035729.1 | - | 17 | 2694 | 5723849 | |
Z69881.1 | - | 17 | 2572 |