Gene Transcripts
- Gene Symbol
- RBM5
- Entrez Gene ID
- 10181
- Gene Name
- RNA binding motif protein 5
- Gene Aliases
- G15, H37, LUCA15, RMB5
- Chromosome Location
- Chr. 3: 50088908 - 50118964 on Build GRCh38
- UniGene ID
- Hs.439480
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005778.3 |
NP_005769.1
|
4, 5 |
518 |
|
NR_036627.2 |
|
4, 5 |
518 |
|
XM_006712917.1 |
XP_006712980.1
|
4, 5 |
518 |
|
XM_011533261.1 |
XP_011531563.1
|
4, 5 |
518 |
|
XM_011533262.1 |
XP_011531564.1
|
4, 5 |
518 |
|
XM_017005503.1 |
XP_016860992.1
|
4, 5 |
500 |
|
XM_017005504.1 |
XP_016860993.1
|
4, 5 |
518 |
|
XM_017005505.1 |
XP_016860994.1
|
4, 5 |
472 |
|
XM_017005506.1 |
XP_016860995.1
|
3, 4 |
396 |
|
XM_017005507.1 |
XP_016860996.1
|
3, 4 |
396 |
|
GenBank mRNA |
AB208813.1 |
- |
4, 5 |
463 |
|
AB586690.1 |
- |
4, 5 |
449 |
|
AF091263.1 |
- |
4, 5 |
491 |
|
AF103802.4 |
- |
4, 5 |
469 |
|
AF107493.1 |
- |
4, 5 |
467 |
|
AK297249.1 |
- |
4, 5 |
472 |
|
AK297445.1 |
- |
4, 5 |
373 |
|
AK302766.1 |
- |
3, 4 |
326 |
|
AK314032.1 |
- |
4, 5 |
473 |
|
BC002957.1 |
- |
4, 5 |
464 |
|
DB080737.1 |
- |
4, 5 |
518 |
|
KU178505.1 |
- |
3, 4 |
343 |
|
KU178506.1 |
- |
3, 4 |
343 |
|
U23946.1 |
- |
4, 5 |
412 |
|
View Full Product Details