Gene Transcripts

Gene Symbol
BPIFB6
Entrez Gene ID
128859
Gene Name
BPI fold containing family B member 6
Gene Aliases
BPIL3, LPLUNC6
Chromosome Location
Chr. 20: 33031648 - 33044108 on Build GRCh38
UniGene ID
Hs.375090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174897.2 NP_777557.1 8, 9 845
XM_017027663.1 XP_016883152.1 8, 9 893
GenBank mRNA AF465767.1 - 8, 9 845

View Full Product Details