Gene Transcripts

Gene Symbol
ROBO1
Entrez Gene ID
6091
Gene Name
roundabout guidance receptor 1
Gene Aliases
DUTT1, SAX3
Chromosome Location
Chr. 3: 78597238 - 79767998 on Build GRCh38
UniGene ID
Hs.744218
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145845.1 NP_001139317.1 2 1078
NM_002941.3 NP_002932.1 4 405
NM_133631.3 NP_598334.2 2 1078
XM_006713277.2 XP_006713340.1 2 1047
XM_011533976.1 XP_011532278.1 4 494
XM_011533977.2 XP_011532279.1 4 347
XM_011533978.1 XP_011532280.1 4 494
XM_011533979.1 XP_011532281.1 4 494
XM_011533980.1 XP_011532282.1 4 494
XM_017006982.1 XP_016862471.1 3 406
XM_017006983.1 XP_016862472.1 2 1047
XM_017006984.1 XP_016862473.1 4 494
XM_017006985.1 XP_016862474.1 2 147
GenBank mRNA AF040990.1 - 3 197
AK297481.1 - 3 353
BC112336.1 - 2 131 40037251
BC115020.1 - 2 179 7961997
BC115022.1 - 2 179 7962000
BC157861.1 - 2 375 9057080
BC171855.1 - 2 375 9054509

View Full Product Details