Gene Transcripts

Gene Symbol
HSPB1
Entrez Gene ID
3315
Gene Name
heat shock protein family B (small) member 1
Gene Aliases
CMT2F, HEL-S-102, HMN2B, HS.76067, HSP27, HSP28, Hsp25, SRP27
Chromosome Location
Chr. 7: 76302558 - 76304297 on Build GRCh38
UniGene ID
Hs.520973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001540.3 NP_001531.1 3 885
GenBank mRNA AK311446.1 - 3 517
BC012768.2 - 3 752 3689220
FJ224323.1 - 3 837
U90906.1 - 3 837

View Full Product Details