Gene Transcripts

Gene Symbol
SIDT2
Entrez Gene ID
51092
Gene Name
SID1 transmembrane family member 2
Gene Aliases
CGI-40
Chromosome Location
Chr. 11: 117179223 - 117197445 on Build GRCh38
UniGene ID
Hs.712144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040455.1 NP_001035545.1 6 694
GenBank mRNA AF151799.1 - 6 736
AK292501.1 - 6 1174
AK292974.1 - 6 685
AK303023.1 - 6 650
AY358442.1 - 6 703
BC114522.1 - 6 723 40035836
KC109742.1 - 6 650

View Full Product Details