Gene Transcripts

Gene Symbol
CLEC1A
Entrez Gene ID
51267
Gene Name
C-type lectin domain family 1 member A
Gene Aliases
CLEC-1, CLEC1
Chromosome Location
Chr. 12: 10069504 - 10099065 on Build GRCh38
UniGene ID
Hs.29549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297748.1 NP_001284677.1 5 1071
NM_001297749.1 NP_001284678.1 4 894
NM_001297750.1 NP_001284679.1 7 1268
NM_001297751.1 NP_001284680.1 6 1212
NM_016511.3 NP_057595.2 6 1170
XM_011520687.2 XP_011518989.1 7 1251
GenBank mRNA AF200949.1 - 6 1108
AK075114.1 - 6 1090
AK091989.1 - 5 1022
AK300979.1 - 4 781
AY358587.1 - 6 1170
BC039072.1 - 6 1076 5178017
BC067746.1 - 6 1090 30337675

View Full Product Details