Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017619.3 | NP_060089.1 | 8, 9 | 1016 | |
GenBank mRNA | AK000015.1 | - | 2, 3 | 191 | |
AK057799.1 | - | 8, 9 | 995 | ||
AY099329.1 | - | 8, 9 | 1001 | ||
BC010697.1 | - | 8, 9 | 992 | 3873751 |