Gene Transcripts

Gene Symbol
BEND5
Entrez Gene ID
79656
Gene Name
BEN domain containing 5
Gene Aliases
C1orf165
Chromosome Location
Chr. 1: 48727523 - 48778603 on Build GRCh38
UniGene ID
Hs.475348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302082.1 NP_001289011.1 5 1181
NM_024603.3 NP_078879.2 5 1099
XM_011542141.2 XP_011540443.1 5 3599
XM_011542142.2 XP_011540444.1 4 1047
XM_017002331.1 XP_016857820.1 6 1132
XM_017002332.1 XP_016857821.1 6 1009
XM_017002334.1 XP_016857823.1 6 1228
GenBank mRNA AK021650.1 - 5 1043
BC007932.2 - 5 1051 4121760
BC011804.2 - 5 865 4336159
KU178776.1 - 5 961
KU178777.1 - 5 1043
KU178778.1 - 4 827

View Full Product Details