Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278651.1 | NP_001265580.1 | 9 | 1568 | |
NM_001317930.1 | NP_001304859.1 | 8 | 1022 | ||
NM_002883.3 | NP_002874.1 | 8 | 1107 | ||
XM_005261695.1 | XP_005261752.1 | 8 | 1235 | ||
XM_005261696.1 | XP_005261753.1 | 8 | 1006 | ||
XM_006724289.3 | XP_006724352.1 | 8 | 1010 | ||
XM_011530293.1 | XP_011528595.1 | 7 | 1100 | ||
XM_011530294.2 | XP_011528596.1 | 8 | 2716 | ||
XM_011530295.2 | XP_011528597.1 | 8 | 1415 | ||
XM_011530297.1 | XP_011528599.1 | 8 | 1235 | ||
XM_017028893.1 | XP_016884382.1 | 9 | 2346 | ||
XM_017028894.1 | XP_016884383.1 | 9 | 2484 | ||
XM_017028895.1 | XP_016884384.1 | 8 | 2846 | ||
XM_017028896.1 | XP_016884385.1 | 9 | 1234 | ||
XM_017028897.1 | XP_016884386.1 | 8 | 2326 | ||
XM_017028898.1 | XP_016884387.1 | 10 | 2259 | ||
XM_017028899.1 | XP_016884388.1 | 7 | 972 | ||
GenBank mRNA | AB058738.1 | - | 7 | 2291 | |
AK302222.1 | - | 7 | 671 | ||
AK303524.1 | - | 8 | 1030 | ||
BC014044.2 | - | 8 | 1022 | 3506039 | |
BC041396.2 | - | 9 | 1578 | 5277434 | |
CR456557.1 | - | 7 | 864 | ||
X82260.1 | - | 7 | 803 |