Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 155018513 - 155033781 on Build GRCh38
UniGene ID
Hs.591491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144622.2 NP_653223.2 9 1578
XM_011509188.2 XP_011507490.1 7 1041
XM_011509189.2 XP_011507491.1 4 1430
GenBank mRNA AK057496.1 - 9 1578
AK097468.1 - 7 992
AK097993.1 - 4 1250
BC111794.1 - 9 1557

View Full Product Details