Gene Transcripts

Gene Symbol
DCST1
Entrez Gene ID
149095
Gene Name
DC-STAMP domain containing 1
Gene Aliases
Chromosome Location
Chr. 1: 155033806 - 155050930 on Build GRCh38
UniGene ID
Hs.567717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143687.2 NP_001137159.1 8 978
NM_152494.3 NP_689707.2 9 1053
GenBank mRNA AK057347.1 - 9 1035
AK301876.1 - 8 955
AK301902.1 - 9 1132
AK301935.1 - 8 764
BC064844.1 - 8 1008 30378702

View Full Product Details