Gene Transcripts

Gene Symbol
SYVN1
Entrez Gene ID
84447
Gene Name
synoviolin 1
Gene Aliases
DER3, HRD1
Chromosome Location
Chr. 11: 65127279 - 65135178 on Build GRCh38
UniGene ID
Hs.75859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032431.2 NP_115807.1 16 2288
NM_172230.2 NP_757385.1 16 2291
GenBank mRNA AB058713.1 - 14 2083
AK074148.1 - 12 3789
AK126912.1 - 16 2291
AL834262.1 - 16 2288
BC013860.1 - 1 322
BC030530.1 - 16 2232 5211742
BC035473.1 - 1 322

View Full Product Details