Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136193.1 | NP_001129665.1 | 2 | 901 | |
NM_001136194.1 | NP_001129666.1 | 2 | 916 | ||
NM_014929.3 | NP_055744.2 | 2 | 808 | ||
GenBank mRNA | AB023188.1 | - | 1 | 549 | |
AF223470.1 | - | 2 | 675 | ||
AK001516.1 | - | 2 | 588 | ||
AK021557.1 | - | 2 | 702 | ||
AL833877.1 | - | 2 | 582 | ||
BC001544.1 | - | 1 | 548 | 3454628 |