Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015697.7 | NP_056512.5 | 2 | 534 | |
XM_011531855.2 | XP_011530157.1 | 2 | 745 | ||
XM_011531857.2 | XP_011530159.1 | 2 | 743 | ||
XM_011531859.2 | XP_011530161.1 | 2 | 742 | ||
XM_011531860.2 | XP_011530162.1 | 2 | 742 | ||
XM_011531862.2 | XP_011530164.1 | 2 | 741 | ||
XM_011531863.2 | XP_011530165.1 | 2 | 738 | ||
XM_011531866.2 | XP_011530168.1 | 2 | 734 | ||
XM_011531867.2 | XP_011530169.1 | 2 | 657 | ||
XM_017008031.1 | XP_016863520.1 | 2 | 657 | ||
GenBank mRNA | AF091086.1 | - | 1 | 84 | |
AJ621061.3 | - | 2 | 534 | ||
AK129583.1 | - | 2 | 290 | ||
AK300972.1 | - | 2 | 415 | ||
BC008804.2 | - | 2 | 391 | 3954787 | |
BC020728.1 | - | 2 | 431 | 4696019 | |
BG699931.1 | - | 2 | 420 |