Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015697.7 | NP_056512.5 | 7 | 1166 | |
XM_017008031.1 | XP_016863520.1 | 7 | 1289 | ||
GenBank mRNA | AF091086.1 | - | 7 | 834 | |
AJ621061.3 | - | 7 | 1166 | ||
AK129583.1 | - | 7 | 922 | ||
BC008804.2 | - | 7 | 1023 | 3954787 | |
BC020728.1 | - | 7 | 1063 | 4696019 | |
BC116454.1 | - | 2 | 210 | ||
BQ363759.1 | - | 4 | 442 |