Gene Transcripts

Gene Symbol
TMEM45A
Entrez Gene ID
55076
Gene Name
transmembrane protein 45A
Gene Aliases
DERP7, DNAPTP4
Chromosome Location
Chr. 3: 100492619 - 100577453 on Build GRCh38
UniGene ID
Hs.658956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018004.2 NP_060474.1 3 662
XM_005247569.2 XP_005247626.1 4 623
XM_017006715.1 XP_016862204.1 4 783
GenBank mRNA AB013911.1 - 2 349
AK000996.1 - 3 662
AK129789.1 - 3 512
AY450392.1 - 3 662
BC040355.1 - 4 596 4748104

View Full Product Details