Gene Transcripts
- Gene Symbol
- TBC1D19
- Entrez Gene ID
- 55296
- Gene Name
- TBC1 domain family member 19
- Gene Aliases
- Chromosome Location
- Chr. 4: 26583924 - 26858919 on Build GRCh38
- UniGene ID
- Hs.479403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001292054.1 |
NP_001278983.1
|
15, 16 |
1401 |
|
NM_018317.3 |
NP_060787.2
|
18, 19 |
1596 |
|
XM_017008363.1 |
XP_016863852.1
|
18, 19 |
1436 |
|
GenBank mRNA |
AB449905.1 |
- |
15, 16 |
1131 |
|
AF075043.1 |
- |
2, 3 |
182 |
|
AK001944.1 |
- |
18, 19 |
1436 |
|
AK225395.1 |
- |
18, 19 |
1436 |
|
AK289631.1 |
- |
18, 19 |
1436 |
|
AK310795.1 |
- |
18, 19 |
1443 |
|
BC031642.1 |
- |
18, 19 |
1392 |
5175317 |
View Full Product Details