Gene Transcripts

Gene Symbol
GPCPD1
Entrez Gene ID
56261
Gene Name
glycerophosphocholine phosphodiesterase 1
Gene Aliases
EDI3, GDE5, GDPD6, PREI4
Chromosome Location
Chr. 20: 5544434 - 5611026 on Build GRCh38
UniGene ID
Hs.636359
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019593.3 NP_062539.1 3, 4 367
XM_005260758.4 XP_005260815.1 4, 5 518
XM_005260761.1 XP_005260818.1 3, 4 367
XM_006723596.3 XP_006723659.1 4, 5 1291
GenBank mRNA AB037855.1 - 3, 4 358
AK308111.1 - 3, 4 365
BC027588.2 - 3, 4 366 4827346
BI561344.1 - 3, 4 300

View Full Product Details