Gene Transcripts

Gene Symbol
ATG9A
Entrez Gene ID
79065
Gene Name
autophagy related 9A
Gene Aliases
APG9L1, MGD3208, mATG9
Chromosome Location
Chr. 2: 219219380 - 219229688 on Build GRCh38
UniGene ID
Hs.323363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077198.2 NP_001070666.1 16 3794
NM_024085.4 NP_076990.4 15 3742
NR_104255.1 16 3735
GenBank mRNA AK021732.1 - 6 1880
AK025822.1 - 10 3096
AK027448.1 - 5 1719
AK056595.1 - 16 3735
AL833865.1 - 9 2991
BC001098.2 - 8 2269
BC021202.2 - 7 2100
BC065534.1 - 16 3673 5787445
BX537984.1 - 15 3663
BX538192.1 - 15 3637
BX538198.1 - 16 3571

View Full Product Details